missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-methylbenzoic acid, 98+%, Thermo Scientific Chemicals
42.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
A building block which is used in preparation of anthranilic acids possessing antibacterial activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A building block which is used in preparation of anthranilic acids possessing antibacterial activity.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
A building block which is used in preparation of anthranilic acids possessing antibacterial activity.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 182°C to 184°C |
Quantity | 5 g |
Beilstein | 2249816 |
Solubility Information | Insoluble in water. |
Formula Weight | 215.05 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-Bromo-2-methylbenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'