missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbenzaldehyde, 95%, Thermo Scientific Chemicals

Catalog Number 15467838
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
24078-12-4
C8H7BrO
199.047
MFCD07787171
RCBPVESMGNZMSG-UHFFFAOYSA-N
4-bromo-2-methyl-benzaldehyde, 2-methyl-4-bromobenzaldehyde, 5-bromo-2-formyltoluene, benzaldehyde, 4-bromo-2-methyl, acmc-1chij, 4-brom-2-methylbenzaldehyd, ksc494k3j, 4-bromo-2-methyl benzaldehyde, benzaldehyde,4-bromo-2-methyl
13555150
4-bromo-2-methylbenzaldehyde
CC1=C(C=CC(=C1)Br)C=O
This item is not returnable. View return policy
24078-12-4
C8H7BrO
199.047
MFCD07787171
RCBPVESMGNZMSG-UHFFFAOYSA-N
4-bromo-2-methyl-benzaldehyde, 2-methyl-4-bromobenzaldehyde, 5-bromo-2-formyltoluene, benzaldehyde, 4-bromo-2-methyl, acmc-1chij, 4-brom-2-methylbenzaldehyd, ksc494k3j, 4-bromo-2-methyl benzaldehyde, benzaldehyde,4-bromo-2-methyl
13555150
4-bromo-2-methylbenzaldehyde
CC1=C(C=CC(=C1)Br)C=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 199.04
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Bromo-2-methylbenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'