missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobiphenyl, 98%, Thermo Scientific™

Catalog Number p-3212020
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
41604-19-7
C12H8BrF
251.10
MFCD00051716
HTRNHWBOBYFTQF-UHFFFAOYSA-N
4-bromo-2-fluorobiphenyl, 4-bromo-2-fluoro-1,1'-biphenyl, 2-fluoro-4-bromo biphenyl, 1,1'-biphenyl, 4-bromo-2-fluoro, 4-bromo-2-fluorodiphenyl, 4-bromo-2-fluoro-1,1-biphenyl, 4-bromo-2-fluoro biphenyl, pubchem2652, acmc-209jkv, 2-fluoro-4-bromobiphenyl
521063
4-bromo-2-fluoro-1-phenylbenzene
FC1=CC(Br)=CC=C1C1=CC=CC=C1
This item is not returnable. View return policy
41604-19-7
C12H8BrF
251.10
MFCD00051716
HTRNHWBOBYFTQF-UHFFFAOYSA-N
4-bromo-2-fluorobiphenyl, 4-bromo-2-fluoro-1,1'-biphenyl, 2-fluoro-4-bromo biphenyl, 1,1'-biphenyl, 4-bromo-2-fluoro, 4-bromo-2-fluorodiphenyl, 4-bromo-2-fluoro-1,1-biphenyl, 4-bromo-2-fluoro biphenyl, pubchem2652, acmc-209jkv, 2-fluoro-4-bromobiphenyl
521063
4-bromo-2-fluoro-1-phenylbenzene
FC1=CC(Br)=CC=C1C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 38°C to 42°C
Color White to Yellow
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula C6H5C6H3(F)Br
Quantity 10 g
Solubility Information Solubility in water: insoluble
Formula Weight 251.1
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Bromo-2-fluorobiphenyl
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-fluorobiphenyl
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'