missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobiphenyl, 98%, ACROS Organics™

Brand:  Acros Organics 373270100

Code : 29

Additional Details : CAS Number : 41604-19-7 Weight : 0.06000kg Transport : UN number : 3152 Chem class : 9 Pack group : II


Quantity 10g
Packaging Glass Bottle

 View more versions of this product

Product Code. 10566023

  • £24.80 / 10g


Estimated Shipment
18-06-2018
Add to basket


Specifications

Chemical Identifiers

4-Bromo-2-fluorobiphenyl
97.5
≥97.5%
C12H8BrF
251.1
41604-19-7
100.0
98%
C6H5C6H3(F)Br
MFCD00051716

Specifications

4-Bromo-2-fluorobiphenyl
97.5
Authentic
Off-white to pale yellow
Crystalline Powder or Chunks
98%
C12H8BrF
251.1
41604-19-7
100.0
Glass Bottle
38° to 42°C
≥97.5%
10g
C6H5C6H3(F)Br
MFCD00051716
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Bromo-2-fluorobiphenyl

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS