missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzyl bromide, 99%, Thermo Scientific™

Catalog Number 10584901
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
76283-09-5
C7H5Br2F
267.92
MFCD00055467
XMHNLZXYPAULDF-UHFFFAOYSA-N
4-bromo-2-fluorobenzyl bromide, 4-bromo-1-bromomethyl-2-fluorobenzene, 2-fluoro-4-bromobenzyl bromide, 4-bromo-2-fluorobenzylbromide, 2-fluoro-4-bromo-benzylbromide, alpha,4-dibromo-2-fluorotoluene, unii-0z26xyc1jy, 4,alpha-dibromo-2-fluorotoluene, 4-bromo-1-bromomethyl-2-fluoro-benzene, 0z26xyc1jy
2733660
4-bromo-1-(bromomethyl)-2-fluorobenzene
C1=CC(=C(C=C1Br)F)CBr
This item is not returnable. View return policy
76283-09-5
C7H5Br2F
267.92
MFCD00055467
XMHNLZXYPAULDF-UHFFFAOYSA-N
4-bromo-2-fluorobenzyl bromide, 4-bromo-1-bromomethyl-2-fluorobenzene, 2-fluoro-4-bromobenzyl bromide, 4-bromo-2-fluorobenzylbromide, 2-fluoro-4-bromo-benzylbromide, alpha,4-dibromo-2-fluorotoluene, unii-0z26xyc1jy, 4,alpha-dibromo-2-fluorotoluene, 4-bromo-1-bromomethyl-2-fluoro-benzene, 0z26xyc1jy
2733660
4-bromo-1-(bromomethyl)-2-fluorobenzene
C1=CC(=C(C=C1Br)F)CBr
CAS Min % 98.5
CAS Max % 100.0
Melting Point >30.0°C
Color White
Boiling Point 126.0°C (19.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 25 g
Formula Weight 267.92
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 4-Bromo-2-fluorobenzyl bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'