missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluoro-N,N-diisobutylbenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H65116.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429386

  • £51.94 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-2-fluoro-N,N-diisobutylbenzenesulfonamide
366.29
MFCD27975720

Specifications

4-Bromo-2-fluoro-N,N-diisobutylbenzenesulfonamide
97%
366.29
Solid
1g
MFCD27975720
Videos
SDS