missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluoro-N-isobutylbenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H65991.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15442679

  • £79.71 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-2-fluoro-N-isobutylbenzenesulfonamide
310.18
MFCD27392033
20408826

Specifications

4-Bromo-2-fluoro-N-isobutylbenzenesulfonamide
Solid
1g
MFCD27392033
149° to 150°C
97%
310.18
20408826
Videos
SDS