missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluoro-N-[4-(trifluoromethyl)phenyl]benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64995.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459306

  • £59.78 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-2-fluoro-N-[4-(trifluoromethyl)phenyl]benzenesulfonamide
398.17
MFCD27975707

Specifications

4-Bromo-2-fluoro-N-[4-(trifluoromethyl)phenyl]benzenesulfonamide
97%
398.17
Solid
1g
MFCD27975707
Videos
SDS