missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluoro-N-(4-methylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64769.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469136

  • £74.73 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-2-fluoro-N-(4-methylphenyl)benzenesulfonamide
344.2
MFCD27824850
4-Bromo-2-fluoro-N-(p-tolyl)benzenesulfonamide

Specifications

4-Bromo-2-fluoro-N-(4-methylphenyl)benzenesulfonamide
97%
344.2
4-Bromo-2-fluoro-N-(p-tolyl)benzenesulfonamide
Solid
1g
MFCD27824850
Videos
SDS