missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluoro-N-(4-fluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64226.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15468786

  • £74.73 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-2-fluoro-N-(4-fluorophenyl)benzenesulfonamide
348.16
MFCD21364211

Specifications

4-Bromo-2-fluoro-N-(4-fluorophenyl)benzenesulfonamide
97%
348.16
Solid
1g
MFCD21364211
Videos
SDS