Alfa Aesar™ 4-Bromo-2-fluoro-N-[4-fluoro-3-(trifluoromethyl)phenyl]benzenesulfonamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H65826.03

1GR 4-Bromo-2-fluoro-N-[4-fluoro-3-(trifluoromethyl)phenyl]benzenesulfonamide, 97% 1g

UNSPSC: 12352105

Code: NEW

Additional Details:
Additional Details: Weight: 0.00100kg


Quantity 1g

Product Code. 15479926

Quantity Price
1 £ 85.44 / 1g
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 4-Bromo-2-fluoro-N-[4-fluoro-3-(trifluoromethyl)phenyl]benzenesulfonamide
Formula Weight 416.16
MDL Number MFCD27975822

Specification

Formula Weight 416.16
Chemical Name or Material 4-Bromo-2-fluoro-N-[4-fluoro-3-(trifluoromethyl)phenyl]benzenesulfonamide
Physical Form Solid
Assay 97%
Quantity 1g
MDL Number MFCD27975822