missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluoro-N-(3-methylbenzyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64474.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458946

  • £99.64 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-2-fluoro-N-(3-methylbenzyl)benzenesulfonamide
358.23
MFCD28098726

Specifications

4-Bromo-2-fluoro-N-(3-methylbenzyl)benzenesulfonamide
97%
358.23
Solid
1g
MFCD28098726
Videos
SDS