missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluoro-N-(3-methoxyphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64027.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15408656

  • £85.44 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-2-fluoro-N-(3-methoxyphenyl)benzenesulfonamide
360.2
MFCD20037277

Specifications

4-Bromo-2-fluoro-N-(3-methoxyphenyl)benzenesulfonamide
Solid
1g
MFCD20037277
Pale brown
97%
360.2
Videos
SDS