missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluoro-N-(3-fluoro-4-methylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64139.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15418726

  • £85.44 / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-2-fluoro-N-(3-fluoro-4-methylphenyl)benzenesulfonamide
362.19
MFCD27975667

Specifications

4-Bromo-2-fluoro-N-(3-fluoro-4-methylphenyl)benzenesulfonamide
97%
362.19
Solid
1g
MFCD27975667
Videos
SDS