missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluoro-N-(2-fluorobenzyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64147.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15418736

  • £66.46 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-2-fluoro-N-(2-fluorobenzyl)benzenesulfonamide
362.19
MFCD27975668

Specifications

4-Bromo-2-fluoro-N-(2-fluorobenzyl)benzenesulfonamide
97%
362.19
Solid
1g
MFCD27975668
Videos
SDS