missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-chloropyrimidine, 98%

Brand:  Alfa Aesar™ H64753.03

Code : NEW

Additional Details : CAS Number : 885702-34-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15452079

  • £238.50 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Description

Description

Applications
Used as an primary and secondary intermediate for pharmaceutical, organic synthesis.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

4-Bromo-2-chloropyrimidine
C4H2BrClN2
MFCD08272196
885702-34-1
193.43
10334333

Specifications

4-Bromo-2-chloropyrimidine
98%
193.43
10334333
885702-34-1
C4H2BrClN2
MFCD08272196
1g
Videos
SDS