missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloroaniline, 99%, Thermo Scientific Chemicals

Catalog Number 10721441
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
38762-41-3
C6H5BrClN
206.47
MFCD00007660
INMZDDDQLHKGPF-UHFFFAOYSA-N
2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
610169
4-bromo-2-chloroaniline
C1=CC(=C(C=C1Br)Cl)N
This item is not returnable. View return policy
38762-41-3
C6H5BrClN
206.47
MFCD00007660
INMZDDDQLHKGPF-UHFFFAOYSA-N
2-chloro-4-bromoaniline, 4-bromo-2-chloro-phenylamine, benzenamine, 4-bromo-2-chloro, 4-bromo-2-chloro-aniline, 4-bromo-2-chlorophenylamine, 4-bromo-6-chloroaniline, 4-bromo-2-chlorobenzenamine, zlchem 546, pubchem3578, 2-chloro4-bromoaniline
610169
4-bromo-2-chloroaniline
C1=CC(=C(C=C1Br)Cl)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 67°C to 70°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H3(Cl)NH2
Quantity 50 g
Beilstein 12, 652
Formula Weight 206.47
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-chloroaniline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-chloroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'