missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2,6-difluorotoluene, 98%

Brand:  Alfa Aesar™ H63807.06

Code : NEW

Additional Details : CAS Number : 179617-08-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15450699

  • £119.78 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-2,6-difluorotoluene
207.02
5-Bromo-1,3-difluoro-2-methylbenzene
179617-08-4
MFCD07368697

Specifications

4-Bromo-2,6-difluorotoluene
Solid
207.02
5-Bromo-1,3-difluoro-2-methylbenzene
179617-08-4
98%
MFCD07368697
5g
Videos
SDS