missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-difluorobenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog Number p-7053878
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
162744-59-4
C7H5BrF2O
223.017
MFCD03094461
LSRHFWSNUFIKER-UHFFFAOYSA-N
4-bromo-2,6-difluorobenzyl alcohol, 4-bromo-2,6-difluorophenyl methanol, 4-bromo-2,6-difluorobenzylalcohol, benzenemethanol, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluoro-benzenemethanol, 2,6-difluoro-4-bromobenzyl alcohol, 4-bromo-2,6-difluoro-phenyl methanol, 2,6-difluoro-4-bromo benzyl alcohol, pubchem4882, acmc-209dox
2773303
(4-bromo-2,6-difluorophenyl)methanol
C1=C(C=C(C(=C1F)CO)F)Br
This item is not returnable. View return policy
162744-59-4
C7H5BrF2O
223.017
MFCD03094461
LSRHFWSNUFIKER-UHFFFAOYSA-N
4-bromo-2,6-difluorobenzyl alcohol, 4-bromo-2,6-difluorophenyl methanol, 4-bromo-2,6-difluorobenzylalcohol, benzenemethanol, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluoro-benzenemethanol, 2,6-difluoro-4-bromobenzyl alcohol, 4-bromo-2,6-difluoro-phenyl methanol, 2,6-difluoro-4-bromo benzyl alcohol, pubchem4882, acmc-209dox
2773303
(4-bromo-2,6-difluorophenyl)methanol
C1=C(C=C(C(=C1F)CO)F)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 76°C to 81°C
Quantity 1 g
Formula Weight 223.01
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Bromo-2,6-difluorobenzyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'