missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-difluorobenzeneboronic acid, 98%, Thermo Scientific™

Catalog Number 15451889
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
352535-81-0
C6H4BBrF2O2
236.81
MFCD03095369
QHYNAULNEIXXPC-UHFFFAOYSA-N
4-bromo-2,6-difluorophenyl boronic acid, 2,6-difluoro-4-bromophenylboronic acid, 4-bromo-2,6-difluorobenzeneboronic acid, boronic acid, b-4-bromo-2,6-difluorophenyl, 4-bromo-2 6-difluorophenylboronic acid, boronic acid,b-4-bromo-2,6-difluorophenyl, pubchem1757, acmc-209if9, 4-bromo-2,6-difluorophenylboronicacid, 4-bromanyl-2,6-bis fluoranyl phenyl boronic acid
4125779
(4-bromo-2,6-difluorophenyl)boronic acid
OB(O)C1=C(F)C=C(Br)C=C1F
This item is not returnable. View return policy
352535-81-0
C6H4BBrF2O2
236.81
MFCD03095369
QHYNAULNEIXXPC-UHFFFAOYSA-N
4-bromo-2,6-difluorophenyl boronic acid, 2,6-difluoro-4-bromophenylboronic acid, 4-bromo-2,6-difluorobenzeneboronic acid, boronic acid, b-4-bromo-2,6-difluorophenyl, 4-bromo-2 6-difluorophenylboronic acid, boronic acid,b-4-bromo-2,6-difluorophenyl, pubchem1757, acmc-209if9, 4-bromo-2,6-difluorophenylboronicacid, 4-bromanyl-2,6-bis fluoranyl phenyl boronic acid
4125779
(4-bromo-2,6-difluorophenyl)boronic acid
OB(O)C1=C(F)C=C(Br)C=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 155°C to 160°C (literature)
Quantity 1 g
Formula Weight 236.81
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-2,6-difluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'