missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,3-difluorobenzaldehyde, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 66°C to 67°C |
Color | White |
Quantity | 250 mg |
Beilstein | 9551272 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 221 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 4-Bromo-2,3-difluorobenzaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'