missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,3-difluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 15481069
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
644985-24-0
C7H3BrF2O
221.001
MFCD09258953
IMOLPSNRLZLWQR-UHFFFAOYSA-N
2,3-difluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2,3-difluoro, 1-bromo-2,3-difluoro-4-formylbenzene, pubchem10068, 4-bromo-2,3-difluoro-benzaldehyde, 4-bromanyl-2,3-bis fluoranyl benzaldehyde, 4-bromo-2, 3-difluorobenzaldehyde
26986390
4-bromo-2,3-difluorobenzaldehyde
C1=CC(=C(C(=C1C=O)F)F)Br
This item is not returnable. View return policy
644985-24-0
C7H3BrF2O
221.001
MFCD09258953
IMOLPSNRLZLWQR-UHFFFAOYSA-N
2,3-difluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2,3-difluoro, 1-bromo-2,3-difluoro-4-formylbenzene, pubchem10068, 4-bromo-2,3-difluoro-benzaldehyde, 4-bromanyl-2,3-bis fluoranyl benzaldehyde, 4-bromo-2, 3-difluorobenzaldehyde
26986390
4-bromo-2,3-difluorobenzaldehyde
C1=CC(=C(C(=C1C=O)F)F)Br

4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 66°C to 67°C
Color White
Quantity 250 mg
Beilstein 9551272
Solubility Information Slightly soluble in water.
Formula Weight 221
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Bromo-2,3-difluorobenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'