missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-1H-indazole, ≥97%

Brand:  Alfa Aesar™ H64527.03

Code : NEW

Additional Details : CAS Number : 186407-74-9 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15401789

  • £15.27 / 1g

Estimated Shipment: 02-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

186407-74-9
197.035
KJIODOACRIRBPB-UHFFFAOYSA-N
22352548
C1=CC2=C(C=NN2)C(=C1)Br
C7H5BrN2
MFCD05664001
4-bromoindazole, 1h-indazole, 4-bromo, 4-bromo 1h indazole, 4-bromo-indazole, bromo-1h-indazole, pubchem7834, 4-bromo-2h-indazole, acmc-1bzds, 4-bromanyl-1h-indazole
4-bromo-1H-indazole

Specifications

4-Bromo-1H-indazole
164°C to 169°C
186407-74-9
MFCD05664001
9757929
KJIODOACRIRBPB-UHFFFAOYSA-N
4-bromo-1H-indazole
22352548
Solid
White
1g
C7H5BrN2
UN2811
4-bromoindazole, 1h-indazole, 4-bromo, 4-bromo 1h indazole, 4-bromo-indazole, bromo-1h-indazole, pubchem7834, 4-bromo-2h-indazole, acmc-1bzds, 4-bromanyl-1h-indazole
C1=CC2=C(C=NN2)C(=C1)Br
197.035
197.04
≥97%
Videos
SDS Documents
Certificates