Learn More
Alfa Aesar™ 4-Bromo-1-methyl-1H-pyrazole, ≥98%
Brand: Alfa Aesar™ H64768.14
Code : NEW
Additional Details : CAS Number : 15803-02-8 Weight : 0.02500kg
Quantity | 25g |
---|
Chemical Identifiers
4-Bromo-1-methyl-1H-pyrazole | |
161 | |
108120 |
15803-02-8 | |
MFCD02179565 |
Specifications
4-Bromo-1-methyl-1H-pyrazole | |
1.5310 | |
185° to 188°C | |
Liquid | |
Colorless to pale yellow | |
MFCD02179565 |
15803-02-8 | |
1.558g/mL | |
25g | |
≥98% | |
161 | |
108120 |