missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-indole, ≥97%, Maybridge

Brand:  Maybridge CC45810DA

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 590417-55-3 Weight : 0.05100kg

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10512774

  • £74.00 / 1g


Estimated Shipment date
from Supplier 31-08-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

4-Bromo-1-methyl-1H-indole
96.5
97%
C9H8BrN
MFCD08435875
590417-55-3
100.0
97%
210.07

Specifications

4-Bromo-1-methyl-1H-indole
96.5
Amber Glass Bottle
97%
C9H8BrN
MFCD08435875
590417-55-3
100.0
1g
97%
210.07
Documentation

Technical Documents