missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-1-butene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
50 g
Description
4-Bromo-1-butene is used in the preparation of 4-(3-butenyloxy)benzoic acid and largazole. It is also used in double alkylation of lactams followed by Grubbs ring-closing metathesis to yield gymnodimine. Further, it serves as a reagent in the preparation of dienehydrazides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromo-1-butene is used in the preparation of 4-(3-butenyloxy)benzoic acid and largazole. It is also used in double alkylation of lactams followed by Grubbs ring-closing metathesis to yield gymnodimine. Further, it serves as a reagent in the preparation of dienehydrazides.
Solubility
Miscible with water, benzene, ether and ethanol.
Notes
Light sensitive. Store in a cool place. Incompatible with strong bases.
4-Bromo-1-butene is used in the preparation of 4-(3-butenyloxy)benzoic acid and largazole. It is also used in double alkylation of lactams followed by Grubbs ring-closing metathesis to yield gymnodimine. Further, it serves as a reagent in the preparation of dienehydrazides.
Solubility
Miscible with water, benzene, ether and ethanol.
Notes
Light sensitive. Store in a cool place. Incompatible with strong bases.
Specifications
Density | 1.33 |
Boiling Point | 98°C to 100°C |
Flash Point | 1°C (34°F) |
Refractive Index | 1.463 |
Quantity | 5 g |
UN Number | UN1993 |
Beilstein | 1098377 |
Sensitivity | Light sensitive |
Solubility Information | Miscible with water,benzene,ether and ethanol. |
SMILES | C=CCCBr |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'