missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-butanol, +85%, ACROS Organics™

Brand:  Acros Organics 390010250

Code : 29

Additional Details : CAS Number : 33036-62-3 Weight : 0.07500kg Transport : UN number : 1987 Chem class : 3 Pack group : II


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10465465

  • £369.40 / 25g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

4-Bromo-1-butanol
85.0
≥85%
C4H9BrO
153.02
33036-62-3
100.0
85+%
Br(CH2)4OH
MFCD00015387

Specifications

4-Bromo-1-butanol
Authentic
100.0
1.0680g/mL
56.0° to 58.0°C (2.0 mmHg)
7°C
≥85%
C4H9BrO
153.02
33036-62-3
85.0
Glass bottle
Clear colorless to brown
25g
Liquid
85+%
Br(CH2)4OH
MFCD00015387
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Bromo-1-butanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS