missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,3-benzodioxole, 97%, Maybridge

£75.00 - £219.00

Chemical Identifiers

CAS 6698-13-1
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD02681890
InChI Key VZPMQHSDFWAZHP-UHFFFAOYSA-N
Synonym 4-bromobenzo d 1,3 dioxole, 4-bromo-2h-1,3-benzodioxole, 4-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 4-bromo, 2,3-methylenedioxy-bromobenzene, 4-bromo-1,3-benzodioxol, methylene dioxy bromo-benzene, 1,3-benzodioxole,4-bromo, 4-bromanyl-1,3-benzodioxole, 1,3-benzodioxol-4-yl bromide
PubChem CID 2776189
IUPAC Name 4-bromo-1,3-benzodioxole
SMILES C1OC2=C(O1)C(=CC=C2)Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10232972 SDS View Documents Maybridge
CC01710CB
250mg Amber Glass Bottle £75.00
250mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 27-02-2020
Please to purchase this item. Need a web account? Register with us today!
10009171 SDS View Documents Maybridge
CC01710DA
1g Amber Glass Bottle £219.00
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 27-02-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

6698-13-1
201.019
VZPMQHSDFWAZHP-UHFFFAOYSA-N
2776189
C1OC2=C(O1)C(=CC=C2)Br
C7H5BrO2
MFCD02681890
4-bromobenzo d 1,3 dioxole, 4-bromo-2h-1,3-benzodioxole, 4-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 4-bromo, 2,3-methylenedioxy-bromobenzene, 4-bromo-1,3-benzodioxol, methylene dioxy bromo-benzene, 1,3-benzodioxole,4-bromo, 4-bromanyl-1,3-benzodioxole, 1,3-benzodioxol-4-yl bromide
4-bromo-1,3-benzodioxole

Specifications

4-Bromo-1, 3-benzodioxole
250mg
96.5
97%
MFCD02681890
VZPMQHSDFWAZHP-UHFFFAOYSA-N
4-bromo-1,3-benzodioxole
2776189
97%
Amber Glass Bottle
6698-13-1
100.0
C7H5BrO2
4-bromobenzo d 1,3 dioxole, 4-bromo-2h-1,3-benzodioxole, 4-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 4-bromo, 2,3-methylenedioxy-bromobenzene, 4-bromo-1,3-benzodioxol, methylene dioxy bromo-benzene, 1,3-benzodioxole,4-bromo, 4-bromanyl-1,3-benzodioxole, 1,3-benzodioxol-4-yl bromide
C1OC2=C(O1)C(=CC=C2)Br
201.019
201.02