missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,2-difluorobenzene, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081789
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
348-61-8
C6H3BrF2
192.99
MFCD00000304
YMQPKONILWWJQG-UHFFFAOYSA-N
1-bromo-3,4-difluorobenzene, 3,4-difluorobromobenzene, benzene, 4-bromo-1,2-difluoro, 3,4-difluorobromebenzene, 4-bromo-1,2-difluoro-benzene, 3,4-difluoro-1-bromobenzene, paragos 390209, fr bf de, pubchem2303, 3,4-difluorophenylbromide
67675
4-bromo-1,2-difluorobenzene
FC1=CC=C(Br)C=C1F
This item is not returnable. View return policy
348-61-8
C6H3BrF2
192.99
MFCD00000304
YMQPKONILWWJQG-UHFFFAOYSA-N
1-bromo-3,4-difluorobenzene, 3,4-difluorobromobenzene, benzene, 4-bromo-1,2-difluoro, 3,4-difluorobromebenzene, 4-bromo-1,2-difluoro-benzene, 3,4-difluoro-1-bromobenzene, paragos 390209, fr bf de, pubchem2303, 3,4-difluorophenylbromide
67675
4-bromo-1,2-difluorobenzene
FC1=CC=C(Br)C=C1F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -4°C
Density 1.707
Boiling Point 150°C to 151°C
Flash Point 33°C (91°F)
Refractive Index 1.505
Quantity 5 g
UN Number UN1993
Beilstein 1934811
Formula Weight 192.99
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-1,2-difluorobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'