missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Boc-amino)indole-3-carboxylic acid, 97%

Brand:  Alfa Aesar™ H63752.03

Code : NEW

Additional Details : CAS Number : 885266-66-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15490599

  • £478.06 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

4-(Boc-amino)indole-3-carboxylic acid
885266-66-0
276.29
MFCD04973970

Specifications

4-(Boc-amino)indole-3-carboxylic acid
Solid
276.29
1g
885266-66-0
97%
MFCD04973970
White
Videos
SDS