missing translation for 'onlineSavingsMsg'
Learn More

4-(BOC-amino)-1-butanol, 98%, ACROS Organics™

£54.50 - £190.00

Chemical Identifiers

CAS 75178-87-9
Molecular Formula C9H19NO3
Molecular Weight (g/mol) 189.25
MDL Number MFCD01861334
InChI Key LIYMTLVBAVHPBU-UHFFFAOYSA-N
Synonym tert-butyl 4-hydroxybutyl carbamate, tert-butyl n-4-hydroxybutyl carbamate, 4-boc-amino-1-butanol, 4-tert-butoxycarbonylamino-1-butanol, carbamic acid, 4-hydroxybutyl-, 1,1-dimethylethyl ester, tert-butyl 4-hydroxybutylcarbamate, n-boc-4-aminobutanol, butylhydroxybutylcarbamate, acmc-1bfe1, 4-t-boc-amino-1-butanol
PubChem CID 545141
IUPAC Name tert-butyl N-(4-hydroxybutyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10109739 SDS View Documents Acros Organics
434210010
1mL Glass bottle £54.50
1mL View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-06-2020
Please to purchase this item. Need a web account? Register with us today!
10047849 SDS View Documents Acros Organics
434210050
5mL Glass bottle £190.00
5mL View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 23-06-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

75178-87-9
189.25
LIYMTLVBAVHPBU-UHFFFAOYSA-N
545141
CC(C)(C)OC(=O)NCCCCO
C9H19NO3
MFCD01861334
tert-butyl 4-hydroxybutyl carbamate, tert-butyl n-4-hydroxybutyl carbamate, 4-boc-amino-1-butanol, 4-tert-butoxycarbonylamino-1-butanol, carbamic acid, 4-hydroxybutyl-, 1,1-dimethylethyl ester, tert-butyl 4-hydroxybutylcarbamate, n-boc-4-aminobutanol, butylhydroxybutylcarbamate, acmc-1bfe1, 4-t-boc-amino-1-butanol
tert-butyl N-(4-hydroxybutyl)carbamate

Specifications

4-(Boc-Amino)-1-butanol
Authentic
1.02
37°C to 40°C
75178-87-9
100.0
C9H19NO3
tert-butyl 4-hydroxybutyl carbamate, tert-butyl n-4-hydroxybutyl carbamate, 4-boc-amino-1-butanol, 4-tert-butoxycarbonylamino-1-butanol, carbamic acid, 4-hydroxybutyl-, 1,1-dimethylethyl ester, tert-butyl 4-hydroxybutylcarbamate, n-boc-4-aminobutanol, butylhydroxybutylcarbamate, acmc-1bfe1, 4-t-boc-amino-1-butanol
CC(C)(C)OC(=O)NCCCCO
189.25
189.25
98%
1.0200g/mL
Glass bottle
Colorless to Yellow
1mL
97.5
97.5% min. (GC)
MFCD01861334
LIYMTLVBAVHPBU-UHFFFAOYSA-N
tert-butyl N-(4-hydroxybutyl)carbamate
545141
Liquid or Low Melting Solid
Safety and Handling

Safety and Handling

Product Identifier
  • 4-(Boc-amino)-1-butanol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.