missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylcarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10696744
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
92-92-2
C13H10O2
198.22
MFCD00002553
NNJMFJSKMRYHSR-UHFFFAOYSA-N
4-biphenylcarboxylic acid, biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, p-phenylbenzoic acid, 4-carboxybiphenyl, para phenyl benzoic acid, 4-diphenylcarboxylic acid, diphenyl-4-carboxylic acid, 4-carboxy-1,1'-biphenyl
66724
4-phenylbenzoic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O
This item is not returnable. View return policy
92-92-2
C13H10O2
198.22
MFCD00002553
NNJMFJSKMRYHSR-UHFFFAOYSA-N
4-biphenylcarboxylic acid, biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, p-phenylbenzoic acid, 4-carboxybiphenyl, para phenyl benzoic acid, 4-diphenylcarboxylic acid, diphenyl-4-carboxylic acid, 4-carboxy-1,1'-biphenyl
66724
4-phenylbenzoic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 223°C to 228°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5C6H4CO2H
Quantity 500 g
Beilstein 09, 671
Formula Weight 198.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Biphenylcarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Biphenylcarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'