missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylpyridine, 97%, ACROS Organics™

£11.84 - £11.94

Chemical Identifiers

CAS 2116-65-6
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006443
InChI Key DBOLXXRVIFGDTI-UHFFFAOYSA-N
Synonym pyridine, 4-phenylmethyl, pyridine, 4-benzyl, 4-benzyl-pyridine, gamma-benzylpyridine, .gamma.-benzylpyridine, 4-phenylmethyl pyridine, unii-1p29gqm0oh, 1p29gqm0oh, 4-benzyl pyridine, 4-benzylpyridine
PubChem CID 16458
IUPAC Name 4-benzylpyridine
SMILES C(C1=CC=CC=C1)C1=CC=NC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10010610
SDS View Documents
Acros Organics
106160050
5mL Glass bottle
£11.94
5mL
View alternative products
Save Up to 
»
In stock, ready to ship: 4
Please to purchase this item. Need a web account? Register with us today!
10665902
SDS View Documents
Acros Organics
106161000
100mL Glass bottle
£11.84
100mL
View alternative products
Save Up to 
»
Estimated Shipment: 10-08-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

2116-65-6
169.23
DBOLXXRVIFGDTI-UHFFFAOYSA-N
16458
C(C1=CC=CC=C1)C1=CC=NC=C1
C12H11N
MFCD00006443
pyridine, 4-phenylmethyl, pyridine, 4-benzyl, 4-benzyl-pyridine, gamma-benzylpyridine, .gamma.-benzylpyridine, 4-phenylmethyl pyridine, unii-1p29gqm0oh, 1p29gqm0oh, 4-benzyl pyridine, 4-benzylpyridine
4-benzylpyridine

Specifications

4-Benzylpyridine
115°C
Glass bottle
1.06
Yellow
5mL
2116-65-6
100.0
C12H11N
MFCD00006443
pyridine, 4-phenylmethyl, pyridine, 4-benzyl, 4-benzyl-pyridine, gamma-benzylpyridine, .gamma.-benzylpyridine, 4-phenylmethyl pyridine, unii-1p29gqm0oh, 1p29gqm0oh, 4-benzyl pyridine, 4-benzylpyridine
DBOLXXRVIFGDTI-UHFFFAOYSA-N
4-benzylpyridine
16458
Liquid
1.0600g/mL
Authentic
1.5810 to 1.5830
287.0°C
9.0°C to 11.0°C
97%
96.0
96% min. (GC)
C6H5CH2(C5H4N)
20, 426
Solubility in water: insoluble in water. Other solubilities: soluble in organic solvents,soluble in ethanol and ether
C(C1=CC=CC=C1)C1=CC=NC=C1
169.23
169.23
97%
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Benzylpyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Product Certifications