missing translation for 'onlineSavingsMsg'
Learn More

(4-Benzyloxyphenyl)acetic acid, 99%, Thermo Scientific™

Catalog Number p-3598216
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6547-53-1
C15H14O3
242.27
MFCD00017540
XJHGAJLIKDAOPE-UHFFFAOYSA-N
4-benzyloxyphenylacetic acid, 2-4-benzyloxy phenyl acetic acid, 4-benzyloxy phenyl acetic acid, 4-benzyloxyphenyl acetic acid, benzeneacetic acid, 4-phenylmethoxy, 4-benzyloxy phenylacetic acid, 4-benzyloxy-phenyl-acetic acid, 2-4-phenylmethoxyphenyl acetic acid, 2-4-phenylmethoxy phenyl acetic acid
81033
2-(4-phenylmethoxyphenyl)acetic acid
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(=O)O
This item is not returnable. View return policy
6547-53-1
C15H14O3
242.27
MFCD00017540
XJHGAJLIKDAOPE-UHFFFAOYSA-N
4-benzyloxyphenylacetic acid, 2-4-benzyloxy phenyl acetic acid, 4-benzyloxy phenyl acetic acid, 4-benzyloxyphenyl acetic acid, benzeneacetic acid, 4-phenylmethoxy, 4-benzyloxy phenylacetic acid, 4-benzyloxy-phenyl-acetic acid, 2-4-phenylmethoxyphenyl acetic acid, 2-4-phenylmethoxy phenyl acetic acid
81033
2-(4-phenylmethoxyphenyl)acetic acid
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(=O)O
CAS Min % 98.5
CAS Max % 100.0
Melting Point 120°C to 122°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4CH2CO2H
Quantity 1 g
Formula Weight 242.27
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material (4-Benzyloxyphenyl)acetic acid
Show More Show Less
compliance-icons
Product Identifier
  • (4-Benzyloxyphenyl)acetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'