missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybromobenzene, 97%, Thermo Scientific Chemicals

Catalog Number p-4813063
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
6793-92-6
C13H11BrO
263.14
OUQSGILAXUXMGI-UHFFFAOYSA-N
4-benzyloxybromobenzene, 1-benzyloxy-4-bromobenzene, 1-bromo-4-phenylmethoxy benzene, 1-bromo-4-benzyloxybenzene, 4-benzyloxy bromobenzene, 1-benzyloxy-4-bromo-benzene, 1-bromo-4-phenylmethyl oxy benzene, 4-benzyloxy-bromobenzene, p-benzyloxy bromobenzene
138835
1-bromo-4-phenylmethoxybenzene
C1=CC=C(C=C1)COC2=CC=C(C=C2)Br
This item is not returnable. View return policy
6793-92-6
C13H11BrO
263.14
OUQSGILAXUXMGI-UHFFFAOYSA-N
4-benzyloxybromobenzene, 1-benzyloxy-4-bromobenzene, 1-bromo-4-phenylmethoxy benzene, 1-bromo-4-benzyloxybenzene, 4-benzyloxy bromobenzene, 1-benzyloxy-4-bromo-benzene, 1-bromo-4-phenylmethyl oxy benzene, 4-benzyloxy-bromobenzene, p-benzyloxy bromobenzene
138835
1-bromo-4-phenylmethoxybenzene
C1=CC=C(C=C1)COC2=CC=C(C=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 62°C to 64°C
Boiling Point 166°C to 167°C (4.0 Torr)
Assay Percent Range 96% min. (GC)
Quantity 10 g
Formula Weight 263.14
Percent Purity 97%
Chemical Name or Material 4-Benzyloxybromobenzene
compliance-icons
Product Identifier
  • 4-Benzyloxybromobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'