missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog Number 10512211
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
836-43-1
C14H14O2
214.26
MFCD00004654
OEBIVOHKFYSBPE-UHFFFAOYSA-N
4-benzyloxybenzyl alcohol, 4-benzyloxy phenyl methanol, p-benzyloxybenzyl alcohol, 4-benzyloxybenzylalcohol, 4-benzyloxy benzyl alcohol, benzenemethanol, 4-phenylmethoxy, p-alkoxybenzyl alcohol resin, 4-phenylmethoxyphenyl methanol, argopore-wang
70043
CHEBI:29486
OCC1=CC=C(OCC2=CC=CC=C2)C=C1
This item is not returnable. View return policy
836-43-1
C14H14O2
214.26
MFCD00004654
OEBIVOHKFYSBPE-UHFFFAOYSA-N
4-benzyloxybenzyl alcohol, 4-benzyloxy phenyl methanol, p-benzyloxybenzyl alcohol, 4-benzyloxybenzylalcohol, 4-benzyloxy benzyl alcohol, benzenemethanol, 4-phenylmethoxy, p-alkoxybenzyl alcohol resin, 4-phenylmethoxyphenyl methanol, argopore-wang
70043
CHEBI:29486
OCC1=CC=C(OCC2=CC=CC=C2)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 84°C to 87°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4CH2OH
Quantity 25 g
IUPAC Name [4-(benzyloxy)phenyl]methanol
Formula Weight 214.26
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Benzyloxybenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzyloxybenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'