missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-2,3-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15497296
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
4-Benzyloxy-2,3-difluorophenylboronic acid pinacol ester
This item is not returnable. View return policy
4-Benzyloxy-2,3-difluorophenylboronic acid pinacol ester

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 346.18
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Benzyloxy-2,3-difluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'