missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-1-butanol, 97%, ACROS Organics™

Brand:  Acros Organics 399120050

Code : 29

Additional Details : CAS Number : 4541-14-4 Weight : 0.06000kg


Quantity 5mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10071504

  • £45.40 / 5mL


Estimated Shipment
06-07-2018
Add to basket


Specifications

Chemical Identifiers

4-Benzyloxy-1-butanol
96.0
97%
C6H5CH2O(CH2)4OH
MFCD00155211
4541-14-4
100.0
C11H16O2
180.25

Specifications

4-Benzyloxy-1-butanol
96.0
Glass bottle
C11H16O2
180.25
5mL
4541-14-4
100.0
97%
C6H5CH2O(CH2)4OH
MFCD00155211
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Benzyloxy-1-butanoL

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS