missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-1-butanol, 97%, Thermo Scientific Chemicals

Catalog Number p-3771224
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
4541-14-4
C11H16O2
180.25
MFCD00155211
TYROJDFHUXSBHC-UHFFFAOYSA-N
4-benzyloxy butan-1-ol, 4-benzyloxy-1-butanol, 1-butanol, 4-phenylmethoxy, 1-butanol, 4-benzyloxy, 4-benyloxybutanol, 4-benzyloxybutanol, 4-benzyloxybutan-1-ol, 4-benzyloxybutyl alcohol, acmc-1agsb
562212
4-phenylmethoxybutan-1-ol
C1=CC=C(C=C1)COCCCCO
This item is not returnable. View return policy
4541-14-4
C11H16O2
180.25
MFCD00155211
TYROJDFHUXSBHC-UHFFFAOYSA-N
4-benzyloxy butan-1-ol, 4-benzyloxy-1-butanol, 1-butanol, 4-phenylmethoxy, 1-butanol, 4-benzyloxy, 4-benyloxybutanol, 4-benzyloxybutanol, 4-benzyloxybutan-1-ol, 4-benzyloxybutyl alcohol, acmc-1agsb
562212
4-phenylmethoxybutan-1-ol
C1=CC=C(C=C1)COCCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Density 1.0260g/mL
Boiling Point 216°C to 256°C
Flash Point 109°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2O(CH2)4OH
Quantity 25 mL
Specific Gravity 1.026
Formula Weight 180.25
Percent Purity 97%
Chemical Name or Material 4-Benzyloxy-1-butanol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzyloxy-1-butanoL
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'