missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-4-hydroxypiperidine, 95%, Thermo Scientific™

Catalog Number p-65481
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
51135-96-7
C12H17NO
191.27
MFCD00023147
KJZBZOFESQSBCV-UHFFFAOYSA-N
4-benzyl-4-hydroxypiperidine, 4-piperidinol, 4-benzyl, 4-benzyl-4-piperidinol, 4-piperidinol, 4-phenylmethyl, acmc-20amoy, pubchem16262, 4benzyl-piperidin-4-ol, maybridge3_002978, 4-benzyl-piperidin-4-ol, 4-benzyl-4-piperidinol #
98152
4-benzylpiperidin-4-ol
C1CNCCC1(CC2=CC=CC=C2)O
This item is not returnable. View return policy
51135-96-7
C12H17NO
191.27
MFCD00023147
KJZBZOFESQSBCV-UHFFFAOYSA-N
4-benzyl-4-hydroxypiperidine, 4-piperidinol, 4-benzyl, 4-benzyl-4-piperidinol, 4-piperidinol, 4-phenylmethyl, acmc-20amoy, pubchem16262, 4benzyl-piperidin-4-ol, maybridge3_002978, 4-benzyl-piperidin-4-ol, 4-benzyl-4-piperidinol #
98152
4-benzylpiperidin-4-ol
C1CNCCC1(CC2=CC=CC=C2)O
CAS Min % 94.0
CAS Max % 100.0
Melting Point 80.0°C to 85.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 191.27
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyl-4-hydroxypiperidine, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzyl-4-hydroxypiperidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'