missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-2-morpholinecarboxylic acid hydrochloride, 97%, Maybridge

£11.00 - £107.00

Chemical Identifiers

CAS 135072-15-0
Molecular Formula C12H16ClNO3
Molecular Weight (g/mol) 257.714
MDL Number MFCD02682031
InChI Key CEDXMALCJZSQHA-UHFFFAOYSA-N
Synonym 4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l
PubChem CID 2776352
IUPAC Name 4-benzylmorpholine-2-carboxylic acid;hydrochloride
SMILES C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10407042 SDS View Documents Maybridge
CC12401CB
250mg £11.00
250mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-07-2020
Please to purchase this item. Need a web account? Register with us today!
10252922 SDS View Documents Maybridge
CC12401DA
1g £34.00
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-07-2020
Please to purchase this item. Need a web account? Register with us today!
10197814 SDS View Documents Maybridge
CC12401DE
5g £107.00
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-07-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

135072-15-0
257.714
CEDXMALCJZSQHA-UHFFFAOYSA-N
2776352
C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl
C12H16ClNO3
MFCD02682031
4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l
4-benzylmorpholine-2-carboxylic acid;hydrochloride

Specifications

4-Benzyl-2-morpholinecarboxylic acid hydrochloride
250mg
97
C12H16ClNO3
4-benzyl-2-morpholinecarboxylic acid hydrochloride, 4-benzylmorpholine-2-carboxylic acid hydrochloride, 4-benzyl-2-carboxymorpholine hydrochloride, 4-benzyl-2-morpholinecarboxylicacidhydrochloride, 4-benzyl-2-morpholinecarboxylic acid hcl, 4-benzyl-morpholine-2-carboxylic acid hydrochloride, 2-morpholinecarboxylic acid, 4-phenylmethyl-, hydrochloride, 2-morpholinecarboxylicacid, 4-phenylmethyl-, hydrochloride 1:1, acmc-1by3p, ksc173i6l
C1COC(CN1CC2=CC=CC=C2)C(=O)O.Cl
257.714
257.72
Amber glass bottle
135072-15-0
97%
MFCD02682031
CEDXMALCJZSQHA-UHFFFAOYSA-N
4-benzylmorpholine-2-carboxylic acid;hydrochloride
2776352
90%