missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzyl-2-morpholinecarboxylic acid hydrochloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 97 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 257.72 |
Percent Purity | 97% |
Chemical Name or Material | 4-Benzyl-2-morpholinecarboxylic acid hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'