missing translation for 'onlineSavingsMsg'
Learn More

(4-Benzyl-1,4-oxazinan-2-yl)methylamine, 97%, Thermo Scientific™

Catalog Number p-2695117
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
110859-47-7
C12H18N2O
206.289
MFCD02682010
CKZVBXBEDDAEFE-UHFFFAOYSA-N
4-benzylmorpholin-2-yl methanamine, 4-benzyl-1,4-oxazinan-2-yl methylamine, 4-benzyl-2-morpholinyl methanamine, 4-benzylmorpholin-2-yl methylamine, 2-morpholinemethanamine, 4-phenylmethyl, 2-aminomethyl-4-benzylmorpholine, 2-morpholinemethanamine,4-phenylmethyl, 1-4-benzylmorpholin-2-yl methanamine, 4-phenylmethyl-2-morpholinyl methanamine
2776360
(4-benzylmorpholin-2-yl)methanamine
C1COC(CN1CC2=CC=CC=C2)CN
This item is not returnable. View return policy
110859-47-7
C12H18N2O
206.289
MFCD02682010
CKZVBXBEDDAEFE-UHFFFAOYSA-N
4-benzylmorpholin-2-yl methanamine, 4-benzyl-1,4-oxazinan-2-yl methylamine, 4-benzyl-2-morpholinyl methanamine, 4-benzylmorpholin-2-yl methylamine, 2-morpholinemethanamine, 4-phenylmethyl, 2-aminomethyl-4-benzylmorpholine, 2-morpholinemethanamine,4-phenylmethyl, 1-4-benzylmorpholin-2-yl methanamine, 4-phenylmethyl-2-morpholinyl methanamine
2776360
(4-benzylmorpholin-2-yl)methanamine
C1COC(CN1CC2=CC=CC=C2)CN
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.29
Percent Purity 97%
Chemical Name or Material (4-Benzyl-1, 4-oxazinan-2-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'