missing translation for 'onlineSavingsMsg'
Learn More

4-Azaindole, 97%, Thermo Scientific Chemicals

Catalog Number p-7055605
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
272-49-1
C7H6N2
118.14
MFCD00971977
XWIYUCRMWCHYJR-UHFFFAOYSA-N
4-azaindole, 1h-pyrrolo 3,2-b pyridine, 1,4-diazaindene, pyrrolopyridine, 4h-pyrrolo 3,2-b pyridine, 1h-pyrrolo 3.2-b pyridine, zlchem 17, pubchem7675, acmc-1clkx
9226
1H-pyrrolo[3,2-b]pyridine
N1C=CC2=NC=CC=C12
This item is not returnable. View return policy
272-49-1
C7H6N2
118.14
MFCD00971977
XWIYUCRMWCHYJR-UHFFFAOYSA-N
4-azaindole, 1h-pyrrolo 3,2-b pyridine, 1,4-diazaindene, pyrrolopyridine, 4h-pyrrolo 3,2-b pyridine, 1h-pyrrolo 3.2-b pyridine, zlchem 17, pubchem7675, acmc-1clkx
9226
1H-pyrrolo[3,2-b]pyridine
N1C=CC2=NC=CC=C12

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in methanol and chloroform. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 125°C to 127°C
Quantity 1 g
Beilstein 109683
Solubility Information Soluble in methanol and chloroform. Slightly soluble in water.
Formula Weight 118.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Azaindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'