missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Azaindole, 97%

Brand:  Alfa Aesar™ H64224.06

Code : NEW

Additional Details : CAS Number : 272-49-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15411329

  • £123.80 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in methanol and chloroform. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

4-Azaindole
118.14
109683
272-49-1
MFCD00971977
Pyrrolo[3,2-b]pyridine

Specifications

4-Azaindole
Solid
118.14
109683
125° to 127°C
272-49-1
97%
MFCD00971977
Pyrrolo[3,2-b]pyridine
5g
Videos
SDS