missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoveratrole, 98%, Thermo Scientific Chemicals

Catalog Number p-58978
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6315-89-5
C8H11NO2
153.18
MFCD00008394
LGDHZCLREKIGKJ-UHFFFAOYSA-N
4-aminoveratrole, benzenamine, 3,4-dimethoxy, 4-aminoveratrol, 3,4-dimethoxybenzenamine, 2-methoxy-4-aminoanisole, 3,4-dimethoxy aniline, aniline, 3,4-dimethoxy, 3,4-dimethyloxy aniline, 3,4-dimethoxy-phenylamine
22770
3,4-dimethoxyaniline
COC1=C(C=C(C=C1)N)OC
This item is not returnable. View return policy
6315-89-5
C8H11NO2
153.18
MFCD00008394
LGDHZCLREKIGKJ-UHFFFAOYSA-N
4-aminoveratrole, benzenamine, 3,4-dimethoxy, 4-aminoveratrol, 3,4-dimethoxybenzenamine, 2-methoxy-4-aminoanisole, 3,4-dimethoxy aniline, aniline, 3,4-dimethoxy, 3,4-dimethyloxy aniline, 3,4-dimethoxy-phenylamine
22770
3,4-dimethoxyaniline
COC1=C(C=C(C=C1)N)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 86°C to 88°C
Color Brown
Boiling Point 174°C to 176°C (22.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC6H3(OCH3)2
Quantity 10 g
Beilstein 13, 780
Solubility Information Solubility in water: insoluble
Formula Weight 153.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminoveratrole, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Aminoveratrole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'