missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyrimidine-5-carbonitrile, 97%, Thermo Scientific™

Catalog Number p-7058896
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
16357-69-0
C5H4N4
120.12
MFCD00054552
JAIYUIOGVNRXEW-UHFFFAOYSA-N
737208
4-aminopyrimidine-5-carbonitrile
NC1=NC=NC=C1C#N
This item is not returnable. View return policy
16357-69-0
C5H4N4
120.12
MFCD00054552
JAIYUIOGVNRXEW-UHFFFAOYSA-N
737208
4-aminopyrimidine-5-carbonitrile
NC1=NC=NC=C1C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 255°C
Quantity 0.25 g
UN Number UN3439
Sensitivity Moisture sensitive
Synonym 4-amino-5-pyrimidinecarbonitrile, 4-amino-pyrimidine-5-carbonitrile, 4-amino-5-cyanopyrimidine, 5-pyrimidinecarbonitrile, 4-amino, pubchem6999, acmc-1busu, 5-pyrimidinecarbonitrile,4-amino, 4-imino-5-cyano-3,4-dihydropyrimidine, 5-pyrimidinecarbonitrile, 4-amino-6ci,7ci,8ci,9ci
Formula Weight 120.11
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Aminopyrimidine-5-carbonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'