missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7082123
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
504-24-5
C5H6N2
94.117
MFCD00006439
NUKYPUAOHBNCPY-UHFFFAOYSA-N
4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine
1727
CHEBI:34385
pyridin-4-amine
C1=CN=CC=C1N
This item is not returnable. View return policy
504-24-5
C5H6N2
94.117
MFCD00006439
NUKYPUAOHBNCPY-UHFFFAOYSA-N
4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine
1727
CHEBI:34385
pyridin-4-amine
C1=CN=CC=C1N

It is used as a research tool, in characterizing subtypes of the potassium channel. It has also been used as a drug, to manage some of the symptoms of multiple sclerosis. In the laboratory, 4-AP is a useful pharmacological tool in studying various potassium conductances in physiology and biophysics. Spinal cord injury patients have also seen improvement with 4-AP therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water(112 g/L at 20 deg C). Soluble in ethyl ether, benzene. Slightly soluble in ligroin. Very soluble in ethanol. Soluble in methanol, acetone, tetrahydrofuran, isopropanol, acetonitrile, N,N-dimethylformamide, dimethylsulfoxide, and ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 157°C to 161°C
Density 1.26
Boiling Point 273°C to 274°C
Flash Point 164°C (327°F)
Odor Unpleasant
Quantity 50 g
UN Number UN2671
Beilstein 105782
Merck Index 14,3933
Solubility Information Soluble in water(112g/L at 20 deg C). Soluble in ethyl ether,benzene. Slightly soluble in ligroin. Very soluble in ethanol. Soluble in methanol,acetone,tetrahydrofuran,isopropanol,acetonitrile,N,N-dimethylformamide,dimethylsulfoxide,and ethanol.
Formula Weight 94.12
Percent Purity 98%
Chemical Name or Material 4-Aminopyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'