missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyridine-3-carboxaldehyde, 95%, ACROS Organics™

Brand:  Acros Organics 437350050

Code : 29

Additional Details : CAS Number : 42373-30-8 Weight : 0.05500kg

Quantity 5g

 View more versions of this product

Product Code. 11926361

  • £482.80 / 5g


Estimated Shipment
19-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Aminopyridine-3-carboxaldehyde
94.0
95%
C6H6N2O
42373-30-8
100.0
95%
122.13

Specifications

4-Aminopyridine-3-carboxaldehyde
Authentic
100.0
111.2° to 112.7°C
5g
95%
122.13
42373-30-8
94.0
White to light yellow to cream
Powder
95%
C6H6N2O
Videos
SDS