missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophthalic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4089706
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5434-21-9
C8H7NO4
181.15
MFCD00013985
OXSANYRLJHSQEP-UHFFFAOYSA-N
4-aminobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-amino, 4-amino-1,2-benzenedicarboxylic acid, 4-amino phthalic acid, contains 3-aminophthalic acid, pubchem16853, acmc-1arqu, 4-aminophthalic acid, ksc270i4n, ghl.pd_mitscher_leg0.1208
72912
4-aminophthalic acid
C1=CC(=C(C=C1N)C(=O)O)C(=O)O
This item is not returnable. View return policy
5434-21-9
C8H7NO4
181.15
MFCD00013985
OXSANYRLJHSQEP-UHFFFAOYSA-N
4-aminobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-amino, 4-amino-1,2-benzenedicarboxylic acid, 4-amino phthalic acid, contains 3-aminophthalic acid, pubchem16853, acmc-1arqu, 4-aminophthalic acid, ksc270i4n, ghl.pd_mitscher_leg0.1208
72912
4-aminophthalic acid
C1=CC(=C(C=C1N)C(=O)O)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 344°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NH2C6H3(COOH)2
Quantity 1 g
Formula Weight 181.15
Percent Purity 97%
Chemical Name or Material 4-Aminophthalic acid
compliance-icons
Product Identifier
  • 4-Aminophthalic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'